NotI-HF®, Restriction Enzymes, New England Biolabs
101641-172
Small Business Enterprise
:
- Concentration:100000 U/ml
- Description:NotI-HF®
- Size:2500 units
- Enzyme Name:NotI-HF
- Cat. No.:101641-172
- Supplier no.:R3189M
An E.coli strain that carries the cloned and modified NotI gene from Nocardia otitidis-caviarum (ATCC 14630).
High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. NotI HF® is engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.