Order Entry
Puerto Rico
ContactUsLinkComponent
NotI
NotI
Catalog # 101228-960
Small Business Enterprise
CAS Number:  
undefined
NotI
Catalog # 101228-960
Supplier Number:  R0189M
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    50000 units/ml
  • Description:
    NotI
  • Size:
    2500 units
  • Enzyme Name:
    NotI
  • Cat. No.:
    101228-960
  • Supplier no.:
    R0189M

Specifications

About this item

An E.coli strain that carries the cloned NotI gene from Nocardia otitidis-caviarum (ATCC 14630).