ContactUsLinkComponent
NEBuffer 4 , New England Biolabs
NEBuffer 4 , New England Biolabs
Catalog # 101227-464
Small Business Enterprise
CAS Number:  
undefined
NEBuffer 4 , New England Biolabs
Catalog # 101227-464
Supplier Number:  B7004S
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    NEBuffer 4
  • Size:
    6.0 mL
  • Cat. No.:
    101227-464
  • Supplier no.:
    B7004S
Specifications
About this item
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity
The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1, NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer are available.