NEBuffer 1 , New England Biolabs
NEBuffer 1, 6.0 mL
101227-458
: B7001S
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity
The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1, NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer are available.
- Description:NEBuffer 1
- Size:6.0 mL