ContactUsLinkComponent
Micrococcal Nuclease
Micrococcal Nuclease
Catalog # 101417-964
Small Business Enterprise
CAS Number:  
undefined
Micrococcal Nuclease
Catalog # 101417-964
Supplier Number:  M0247S
Small Business Enterprise
CAS Number:  
Specifications
  • Size:
    320000 gel units
  • Enzyme Name:
    Micrococcal Nuclease
  • Cat. No.:
    101417-964
  • Supplier no.:
    M0247S
Specifications
About this item
Micrococcal nuclease is derived from Staphylococcus aureus and is a relatively non-specific endo-exonuclease
It is purified from a recombinant E. coli strain that digests double-stranded, single-stranded, circular and linear nucleic acids. The enzyme is active in the pH range of 7.0 - 10.0, with optimal activity at pH 9.2 for both RNA and DNA substrates. Cleavage preferences have been observed at sites rich in adenylate, deoxyadenylate or thymidylate. Both DNA and RNA are degraded to 3´ phosphomononucleotides and dinucleotides.