ContactUsLinkComponent
M13K07 Helper Phage, New England Biolabs
M13K07 Helper Phage, New England Biolabs
  101228-402
Small Business Enterprise
 :  
undefined
M13K07 Helper Phage, New England Biolabs
  101228-402
 :  N0315S
Small Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    M13K07 Helper Phage
  • Size:
    1.8 mL
  • Cat. No.:
    101228-402
  • Supplier no.:
    N0315S
M13KO7 is an M13 derivative which carries the mutation Met40Ile in gII, with the origin of replication from P15A and the kanamycin resistance gene from Tn903 both inserted within the M13 origin of replication
M13KO7 is able to replicate in the absence of phagemid DNA. In the presence of a phagemid bearing a wild-type M13 or f1 origin, single-stranded phagemid is packaged preferentially and secreted into the culture medium. This allows easy production of single-stranded phagemid DNA for mutagenesis or sequencing.