Order Entry
United States
ContactUsLinkComponent
Human Recombinant Histone H3.1
Human Recombinant Histone H3.1
Catalog # 200067-194
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
undefined
Human Recombinant Histone H3.1
Catalog # 200067-194
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
    Human
  • Size:
    100 μg
  • Environmentally Preferable:
  • Protein/Peptide Name:
    Histone H3.1
  • Cat. No.:
    200067-194
  • Supplier no.:
    M2503S

Specifications

About this item

Histone H3 combines with Histone H4 to form the H3/H4 tetramer. Two H2A/H2B heterodimers interact with an H3/H4 tetramer to form the histone octamer. It is also modified by various enzymes and can act as a substrate for them. These modifications have been shown to be important in gene regulation. Histone H3.1, an H3 variant that has thus far only been found in mammals, is replication dependent and is associated with gene activation and gene silencing.