ContactUsLinkComponent
Gel Loading Dye Orange (6X), New England Biolabs
  101417-882
Small Business Enterprise
 :  
undefined
Gel Loading Dye Orange (6X), New England Biolabs
  101417-882
 :  B7022S
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Gel Loading Dye Orange (6X)
  • Size:
    6.0 mL
  • Environmentally Preferable:
  • Cat. No.:
    101417-882
  • Supplier no.:
    B7022S
Gel Loading Dye, Orange (6X) is a pre-mixed loading buffer with a tracking dye for agarose and non-denaturing polyacrylamide gel electrophoresis
This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to their DNA substrates following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. Orange G will not appear in gel photographs, and runs ahead of all but the smallest restriction fragments. Orange G migrates at approximately 50 bp on a standard 1% TBE agarose gel.