ContactUsLinkComponent
Bst DNA Polymerase Full Length , New England Biolabs
Bst DNA Polymerase Full Length , New England Biolabs
Catalog # 101449-324
Small Business Enterprise
CAS Number:  
undefined
Bst DNA Polymerase Full Length , New England Biolabs
Catalog # 101449-324
Supplier Number:  M0328S
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Bst DNA Polymerase Full Length
  • Size:
    500 units
  • Cat. No.:
    101449-324
  • Supplier no.:
    M0328S
Specifications
About this item
Bst DNA Polymerase, Full Length is the full length polymerase from Bacillus stearothermophilus
It has 5´ → 3´ polymerase and double-strand specific 5´ → 3´ exonuclease activity, but lacks 3´ → 5´ exonuclease activity.