BG-Maleimide, New England Biolabs
Catalog # 101447-710
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Specifications
- Description:BG-Maleimide
- Size:2 mg
- Cat. No.:101447-710
- Supplier no.:S9153S
Specifications
About this item
BG-Maleimide is a thiol-reactive building block for the one-step synthesis of SNAP-tag® substrates from thiol-containing precursors such as thiol-modified oligonucleotides
This building block allows you to make custom SNAP-tag substrates for labeling SNAP-tag fusion proteins for a wide range of applications such as the coupling of proteins and solid surfaces such as DNA chips.