Distilling Receiver, Graduated, with Hooks, Glassco
89426-592
: Glassco
- Capacity:15 ml
- Description:Distilling Receiver, Graduated, with Hooks
- Cat. No.:89426-592
- Top [ST] joint size:[ST] 14/20
- Supplier no.:088.468.03
Centrifuge tube shaped receiver with an upper standard taper joint
Receiver also features two glass hooks 180° apart.