Reflux Condenser, Glassco
Supplier: Glassco
Certificates
About this item
Coil style reflux condenser with tightly wrapped coil providing maximum cooling
Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 8mm or 10mm at the middle serration.