To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:pH test in cooling lubricants
- Size:50 strips
- Cat. No.:EM1.16898.0001
- pH range:7.0 - 10.0
- Supplier no.:1.16898.0001
Specifications
About this item
In water analysis the pH value plays a very important role in how other substances behave. The test can be used for monitoring the pH value in cooling lubricants.
- Store at 15 to 25 °C
Reflectoquant® pH tests are suited for all media in environmental analysis and in industrial in-process controls. The reaction zones of the test strip change color depending on the pH. The responsible indicator is chemically bound to the zones and hence does not bleed. The resulting color is evaluated reflectometrically on the RQflex®.