Reflectoquant® Lactic Acid Test, MilliporeSigma
Catalog # EM1.16127.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Description:Lactic Acid Test Method, Reflectometric with Test Strips
- Cat. No.:EM1.16127.0001
- Range:3.0 - 60.0 mg/L
- Supplier no.:1.16127.0001
- No. of strips:50
Specifications
About this item
The test measures total lactic acid (sum of D- and L-lactic acid in beverages, e.g. wine, beer and fruit/vegetable juice after dilution. Furthermore the test can be used for the determination of lactic acid in food after pretreatment. Enzymatic reaction.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.