ContactUsLinkComponent
Lab Armor® Walkabout Trays, Shel Lab
Lab Armor® Walkabout Trays, Shel Lab
Certificates
undefined
Lab Armor® Walkabout Trays, Shel Lab
Specifications
  • Catalog No:
  • 89409-260
  • 89409-258
  • Capacity:
  • 0.5 – 1.0 L (beads)
  • 0.5 – 1.0 L (beads)
  • Temperature Range:
  • -196 to 93°C
  • -196 to 93°C
  • Dimensions:
  • 15.24 x 15.24 x 7.6 cm (6 x 6 x 3")
  • 15.24 x 15.24 x 7.6 cm (6 x 6 x 3")
Specifications