Order Entry
Puerto Rico
ContactUsLinkComponent
 
HiScreen Capto Adhere Columns, Cytiva
Catalog # 97067-802
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
HiScreen Capto Adhere Columns, Cytiva
Catalog # 97067-802
Supplier:  Cytiva
Supplier Number:  28-9269-81
Product available on GSA Advantage®

Specifications

  • Particle Size:
    75 µm
  • Max. Pressure:
    3 bar [0.3 MPa] (44 psi)
  • Capacity:
    4.7 ml
  • Description:
    HiScreen™ Capto™ adhere
  • pH Range:
    3 - 12
  • Storage:
    4 to 30 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    7.7×100 mm
  • Flow velocity:
    < 600 cm/h
  • Cat. No.:
    97067-802
  • Supplier no.:
    28926981

Specifications

About this item

HiScreen Capto adhere columns are prepacked with Capto adhere ion exchange chromatography resins, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same resin using the same linear fluid velocity

Capto adhere is a multimodal anion exchanger for intermediate purification and polishing of monoclonal antibodies after the protein A capture step, giving the possibility to design a two-step chromatographic downstream process.

The small column volume of 4.7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.