Order Entry
Puerto Rico
ContactUsLinkComponent
HiScreen Q FF Columns
HiScreen Q FF Columns
Catalog # 97067-816
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
HiScreen Q FF Columns
Catalog # 97067-816
Supplier:  Cytiva
Supplier Number:  28-9505-10
Product available on GSA Advantage®

Specifications

  • Particle Size:
    90 µm
  • Max. Pressure:
    1.5 bar [0.15 MPa] (22 psi)
  • Capacity:
    4.7 ml
  • Description:
    HiScreen™ Q FF
  • pH Range:
    2 - 12
  • Storage:
    4...30 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    7.7×100 mm
  • Flow velocity:
    < 450 cm/h
  • Cat. No.:
    97067-816
  • Supplier no.:
    28950510

Specifications

About this item

HiScreen Q FF columns are prepacked with Q Sepharose Fast Flow, a strong anion exchanger, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity

The small column volume of 4.7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

Q Sepharose Fast Flow is a strong anion exchanger and IEX Sepharose Fast Flow medium are well-proven, industrial standards for ion exchange chromatography.