HiTrap IEX Selection Kit, Cytiva
Catalog # 95055-856
Supplier: Cytiva
Product available on GSA Advantage®
Specifications
- Flow rate:< 4 mL/min
- Max. Pressure:5 bar [0.5 MPa] (70 psi)
- Capacity:1 ml
- Environmentally Preferable:
- Description:95055-856
- Storage:4 to 30 °C, 20% Ethanol (Q, DEAE, ANX,CM), 20% Ethanol, 0.2 M Sodium Acetate (SP)
- Bed dimensions:7×25 mm
- Cat. No.:95055-856
- Supplier no.:17-6002-33
Specifications
About this item
HiTrap IEX Selection Kit is used for screening of the optimal Cytiva ion exchange resin for specific application and development work.
- Fast and easy selection: Seven prepacked HiTrap columns, each containing a different Sepharose ion exchange resin (medium)
- Convenient way to screen for the optimal Cytiva anion exchange (aex) or cation exchange (cex) resin for a given application
- Excellent for optimization studies; buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
- HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
Cytiva's HiTrap IEX Selection Kit contains a variety of anion and cation exchange resins, allowing users to screen for the most appropriate column.
Ordering information: Prepacked HiTrap IEX columns are also available individually.
Delivery: The kit includes seven 1 ml HiTrap columns packed with the following ion exchange resins: SP Sepharose Fast Flow cex column (strong cation exchanger), SP Sepharose XL cex column (strong cation exchanger), CM Sepharose Fast Flow cex column (weak cation exchanger), Q Sepharose Fast Flow aex column (strong anion exchanger), Q Sepharose XL aex column (strong anion exchanger), DEAE Sepharose Fast Flow aex column (weak anion exchanger), ANX Sepharose 4 Fast Flow aex column (weak anion exchanger).
Packaging: The ion exchange resins in the columns are offered as bulk resin and in other prepacked formats.