ContactUsLinkComponent
HiTrap Columns, Capto SP ImpRes, Cytiva
HiTrap Columns, Capto SP ImpRes, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, Capto SP ImpRes, Cytiva
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 97067-776
  • 97067-778
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • Storage:
  • 4 to 30 °C, 20% Ethanol, 0.2 M Sodium Acetate
  • 4 to 30 °C, 20% Ethanol, 0.2 M Sodium Acetate
Specifications