Order Entry
Puerto Rico
ContactUsLinkComponent
 
HiTrap Columns, SP Sepharose FF, Cytiva
 :  Cytiva
 
undefined
HiTrap Columns, SP Sepharose FF, Cytiva
 :  Cytiva
 

 

  • Catalog No:
  • 95056-082
  • 95056-106
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • pH Range:
  • 4 - 13
  • 4 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol, 0.2 M Sodium Acetate
  • 4 to 30 °C, 20% Ethanol, 0.2 M Sodium Acetate