ContactUsLinkComponent
HiTrap Columns, Q Sepharose FF, Cytiva
HiTrap Columns, Q Sepharose FF, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, Q Sepharose FF, Cytiva
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 95056-104
  • 95056-080
  • Description:
  • 95056-104
  • 95056-080
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
Specifications