Order Entry
Puerto Rico
ContactUsLinkComponent
 
RESOURCE S Columns, Cytiva
 :  Cytiva
 
undefined
RESOURCE S Columns, Cytiva
 :  Cytiva
 

 

  • Catalog No:
  • 95055-802
  • 95056-190
  • Description:
  • RESOURCE™ S
  • RESOURCE™ S
  • Max. Pressure:
  • 15 bar [1.5 MPa] (218 psi)
  • 6 bar [0.6 MPa] (87 psi)
  • pH Range:
  • 2 - 13
  • 2 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol, 0.2 M Sodium Acetate
  • 4 to 30 °C, 20% Ethanol, 0.2 M Sodium Acetate