Order Entry
Puerto Rico
ContactUsLinkComponent
 
RESOURCE Q Columns, Cytiva
 :  Cytiva
 
undefined
RESOURCE Q Columns, Cytiva
 :  Cytiva
 

 

  • Catalog No:
  • 95056-188
  • 95055-800
  • Description:
  • RESOURCE™ Q
  • RESOURCE™ Q
  • Max. Pressure:
  • 6 bar [0.6 MPa] (87 psi)
  • 15 bar [1.5 MPa] (218 psi)
  • pH Range:
  • 2 - 12
  • 2 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol