HiPrep Sephacryl S-400 HR, Cytiva
: Cytiva
HiPrep™ 16/60 Sephacryl® S-400 HR and HiPrep™ 26/60 Sephacryl® S-400 HR are pre-packed gel filtration columns for preparative separation of biomolecules of up to 5 and 13 ml sample volume respectively. Sephacryl® S-400 is used for separating polysaccarides, macromolecules with extended structures, and even small plasmids.
- Maximum recovery and minimum non specific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Columns provides reproducible separations over a wide molecular weight range
- Catalog No:
- 97067-768
- 97067-770
- Max. Pressure:
- 1.5 bar [0.15 MPa] (22 psi)
- 1.5 bar [0.15 MPa] (22 psi)
- pH Range:
- 3 - 11
- 3 - 11
- Storage:
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol