About this item
GSTrap™ HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins.
- High resolution purification of GST-tagged proteins
- Mild elution conditions preserving protein antigenicity and function
- Fast, one-step purification
- Easy to use with syringe, pump or a chromatography system
GSTrap™ HP 1 and 5 ml columns are prepacked with Glutathione Sepharose High Performance resin, which provides high-resolution purifications, thus a more concentrated sample. GST-tagged proteins are purified from pretreated bacterial lysates by affinity chromatography using immobilized glutathione, such as Glutathione Sepharose High Performance. GST-tagged proteins are bound to the affinity resin, and impurities are removed by washing with binding buffer
GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods.
Specifications
- Catalog No:
- 95056-212
- 95056-020
- 95056-018
- Max. Pressure:
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- pH Range:
- 3 - 12
- 3 - 12
- 3 - 12
- Storage:
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
Specifications
Related Information
Certificates
Search by Lot Number17528101_DATA FILE 1.pdf 17528101_DATA FILE 2.pdf 17528101_DATA FILE.pdf 17528201_DATA FILE 1.pdf 17528201_DATA FILE.pdf
Datasheet
17528101_HAND BOOK 1.pdf 17528101_HAND BOOK 2.pdf 17528101_HAND BOOK.pdf 17528101_INSTRUCTIONS.pdf 17528201_HAND BOOK 1.pdf
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements