Order Entry
Puerto Rico
ContactUsLinkComponent
 
HisTrap Columns, HP
 :  Cytiva
 
undefined
HisTrap Columns, HP
 :  Cytiva
 

 

  • Catalog No:
  • 95055-912
  • 95056-206
  • 89501-388
  • 95056-204
  • Description:
  • HisTrap™ HP
  • HisTrap™ HP
  • HisTrap™ HP
  • HisTrap™ HP
  • Binding Capacity:
  • At least 40 mg histidine-tagged protein/ml medium
  • At least 40 mg histidine-tagged protein/ml medium
  • At least 40 mg histidine-tagged protein/ml medium
  • At least 40 mg histidine-tagged protein/ml medium
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • pH Range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol

 

Frequently Bought Together