Order Entry
Puerto Rico
ContactUsLinkComponent
HiTrap Columns, NHS-Activated HP, Cytiva
HiTrap Columns, NHS-Activated HP, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, NHS-Activated HP, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 95055-952
  • 95055-954
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • pH Range:
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 8 °C, 100% Isopropanol
  • 4 to 8 °C, 100% Isopropanol

Specifications

Frequently Bought Together