Order Entry
Puerto Rico
ContactUsLinkComponent
HiScreen Capto L Columns, Cytiva
HiScreen Capto L Columns, Cytiva
Catalog # 89234-990
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
HiScreen Capto L Columns, Cytiva
Catalog # 89234-990
Supplier:  Cytiva
Supplier Number:  17-5478-14
Product available on GSA Advantage®

Specifications

  • Particle Size:
    85 µm
  • Max. Pressure:
    3 bar [0.3 MPa] (44 psi)
  • Capacity:
    4.7 ml
  • Description:
    HiScreen™ Capto™ L
  • pH Range:
    2 - 10
  • Storage:
    2 to 8 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    7.7×100 mm
  • Cat. No.:
    89234-990
  • Supplier no.:
    17547814

Specifications

About this item

HiScreen™ Capto™ L columns are prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.

  • Prepacked, ready to use columns for convenient process development optimisation
  • Bed height of 10 cm allows method optimisation and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume (4.7 ml) gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity

The columns are an excellent choice for method optimisation and parameter screening.