ContactUsLinkComponent
Streptavidin HP MultiTrap™, Cytiva
Streptavidin HP MultiTrap™, Cytiva
Catalog # 95055-976
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Streptavidin HP MultiTrap™, Cytiva
Catalog # 95055-976
Supplier:  Cytiva
Supplier Number:  28-9031-31
Product available on GSA Advantage®
Specifications
  • Description:
    Streptavidin HP MultiTrap, < 700 x g
  • Cat. No.:
    95055-976
  • Supplier no.:
    28-9031-31
Specifications
About this item
Streptavidin HP MultiTrap can be used for protein enrichment, where a biotinylated antibody (or similar affinity molecule) is attached to the Streptavidin and the protein of interest is enriched through the affinity interaction with the antibody.
  • Fast, reliable protein enrichment through immobilized biotinylated biomolecules in prepacked 96-well filter plates that are ready to use
  • Fast and flexible protocol with elution conditions formatted for both electrophoresis and LC-MS analysis workflows
  • Useful for exploiting either the strong interactions of biotin and streptavidin or the somewhat weaker interaction of 2-iminobiotin and streptavidin
  • Each well is prepacked with Streptavidin Sepharose High Performance for reproducibility and high performance
  • Easy scale-up to Streptavidin HP SpinTrap or HiTrap Streptavidin HP prepacked columns
  • MultiTrap 96-well filter plates can be used with centrifugation or vacuum, manually or in automated robotic systems
Streptavidin HP MultiTrap can also be used for the direct enrichment of proteins that are biotinylated. By binding a specific biotinylated protein to the column, protein-protein interactions can also be investigated.
Ordering information: Collection plates have to be ordered separately.