ContactUsLinkComponent
His MultiTrap TALON Filter Plates, Cytiva
His MultiTrap TALON Filter Plates, Cytiva
Catalog # 11010-722
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
His MultiTrap TALON Filter Plates, Cytiva
Catalog # 11010-722
Supplier:  Cytiva
Supplier Number:  29-0005-96
Product available on GSA Advantage®
Specifications
  • Description:
    His MultiTrap™ TALON®, 96-well plate
  • Cat. No.:
    11010-722
  • Supplier no.:
    29000596
Specifications
About this item
His MultiTrap™ TALON® multiwell plates are designed for high-throughput screening of histidine-tagged proteins from clarified cell lysates. His MultiTrap™ TALON® prepacked plates are manufactured with high reproducibility with regards to the well and media volumes in order to enable consistent results. The average purity of His MultiTrap TALON® is >85% with yields up to 1 mg/well. Leakage of cobalt ions from TALON® Superflow™ medium is negligible. His MultiTrap™ TALON® is used manually with vacuum or centrifugation, or in a liquid handling station.
  • Up to 1 mg of pure histidine-tagged protein per well
  • Large sample volumes can be applied all at once
Prefilled with 50 µl TALON® Superflow™ cobalt-IMAC (immobilized metal affinity chromatography) medium for reproducible and high throughput parallel screening, offering a different selectivity compared to nickel-charged media.