Order Entry
Puerto Rico
ContactUsLinkComponent
PD MiniTrap G-10 Columns, Cytiva
PD MiniTrap G-10 Columns, Cytiva
Catalog # 95056-252
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
PD MiniTrap G-10 Columns, Cytiva
Catalog # 95056-252
Supplier:  Cytiva
Supplier Number:  28-9180-10
Product available on GSA Advantage®

Specifications

  • Capacity:
    2.1 ml
  • Description:
    PD MiniTrap G-10
  • Wetted Materials:
    No
  • Storage:
    4 to 30°C, 20% Ethanol
  • Bed dimensions:
    9.7×28 mm
  • Cat. No.:
    95056-252
  • Sample volume:
    100 - 300 μl
  • Supplier no.:
    28-9180-10

Specifications

About this item

PD MiniTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.

  • Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700 before downstream analysis such as liquid chromatography, electrophoresis, and MS
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels
  • High desalting capacity
  • Preparation of samples in the volume range of 100 to 300 μl

PD MiniTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system. PD MiniTrap G-10 columns are a member of the Trap platform, which addresses the need for flexible, small-scale preparation of proteins and peptides before analysis.

For larger scale please use PD MidiTrap G-10 columns.

Delivery: Instructions are supplied with the columns.