DNA (Deoxyribonucleic acid) (from calf thymus), activated
Catalog # 95040-288
Supplier: Cytiva
Product available on GSA Advantage®
CAS Number:
Specifications
- Size:25 units/ml
- Cat. No.:95040-288
- Supplier no.:27-4575-01
Specifications
About this item
:
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922