Order Entry
Puerto Rico
ContactUsLinkComponent
Accessories for PIG® Spill Kit in Small Wall-Mount Cabinet, New Pig
Accessories for PIG® Spill Kit in Small Wall-Mount Cabinet, New Pig
Catalog # 56620-230
Supplier:  New Pig
undefined
Accessories for PIG® Spill Kit in Small Wall-Mount Cabinet, New Pig
Catalog # 56620-230
Supplier:  New Pig
Supplier Number:  KIT216

Specifications

  • Capacity:
    Refill for PIG® Spill Kit in Small Wall-Mount Cabinet, Absorbs up to 45.4 L (12 gal.)
  • Description:
    Refill for PIG® Spill Kit in Small Wall-Mount Cabinet
  • Cat. No.:
    56620-230
  • Absorbency:
    45.4 L (12 gal.)
  • Supplier no.:
    KIT216
  • Details:
    75× - 10" W x 13" L PIG® Absorbent Mat Pad in Dispenser Box; 10× - 10" W x 10" L x 2" H PIG® Absorbent Pillow; 56× - 11.8" W x 13" L PIG® PR40 All-Purpose Wipers; 5× - 18" W x 30" H Polyethylene Disposal Bags (BAG201-S)

Specifications

About this item

Refill for PIG® Spill Kit in Small Wall-Mount Cabinet. Don't be caught with an empty spill kit when you need it most! Keep a refill handy to instantly replace used contents.

  • Packed to replace used absorbents from your PIG Spill Kit in Small Wall-Mount Cabinet (KIT215)
  • PIG Mat and Pillows absorb; wipers clean surfaces and hands
  • Temporary disposal bags with ties help make cleanup faster

Only PIG Spill Kit Refills feature PIG Absorbents proven for rapid response.