ContactUsLinkComponent
PIG® HazMat Spill Kit in 95-Gallon High-Visibility Container, New Pig
PIG® HazMat Spill Kit in 95-Gallon High-Visibility Container, New Pig
Catalog # 56620-434
Supplier:  New Pig
undefined
PIG® HazMat Spill Kit in 95-Gallon High-Visibility Container, New Pig
Catalog # 56620-434
Supplier:  New Pig
Supplier Number:  KIT362
Specifications
  • Capacity:
    Up to 238.5 L (63 gal.)
  • Environmentally Preferable:
  • Cat. No.:
    56620-434
  • Supplier no.:
    KIT362
Specifications
About this item
Lift-out, prepacked baskets speed access and guard contents from UV
Lime green drum is easy to locate. Lightweight, 100% polyethylene drum resists chemicals and keeps contents clean and dry. Large, color-coded label makes identification fast and accurate. Easy-access threaded lid (not rated for shipping). Tamperproof seal helps prevent pilfering of spill response supplies.
No wheels. Not UN rated.
PIG HazMat Socks and Dikes stop spreading spills; PIG HazMat Pads and Pillows absorb quickly. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide