- Capacity:Up to 117.3 L (31 gal.)
- Cat. No.:89205-846
- Supplier no.:KIT468
Overpack drum container is UN Rated for shipping waste after oil spill cleanup
Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies. UN1H2/X125/S.
The 6" olefin wheels make spill kit highly mobile for speedy response anywhere in your facility. Lift-out, prepacked baskets speed access and guard contents from UV. PIG Oil-Only Booms stop spreading spills; PIG Stat-Mat and Pillows absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables