Order Entry
Puerto Rico
ContactUsLinkComponent
PIG® Spill Kit in Large Mobile Container, New Pig
PIG® Spill Kit in Large Mobile Container, New Pig
Catalog # 56620-362
Supplier:  New Pig
undefined
PIG® Spill Kit in Large Mobile Container, New Pig
Catalog # 56620-362
Supplier:  New Pig
Supplier Number:  KIT259

Specifications

  • Capacity:
    Up to 140 L (37 gal.)
  • Cat. No.:
    56620-362
  • Supplier no.:
    KIT259

Specifications

About this item

Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry

  • Prepacked container provides easy access to contents
  • Built-in handles allow easy steering through tight spots
  • Large, color-coded label makes identification fast and accurate.
  • Hinged lid provides easy access
  • Temporary disposal bags help make cleanup easier
  • Tamperproof seal helps prevent pilfering of spill response supplies

8" rubber wheels. Not UN rated.

Heavy-duty 8" rubber wheels roll easily over nearly any surface. Equipped with PIG Lite-Dri and Shop Dri Loose Absorbents along with PIG Blue Absorbent Socks, Mat and Pillows for spill response on rough surfaces.