Specifications
- Capacity:Up to 113.6 L (30 gal.)
- Cat. No.:89094-558
- Supplier no.:KIT272
Specifications
About this item
Overpack drum container is UN Rated for shipping waste after spill cleanup
Temporary disposal bags help make cleanup easier. Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies.
6" polyolefin wheels. UN1H2/X125/S.
Lift-out, prepacked baskets speed access and guard contents from UV. PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb; wipers clean surfaces and hands.