- Description:Single edge razor blade pop-up dispenser incl. 100 blades
- Environmentally Preferable:
- Cat. No.:95062-667
- Supplier no.:62-0325
The Pop-up dispenser enables the user to access a blade without having to touch it before use.
Simple operation allows the blade to be unwrapped and made accesible with the push of a button. This dispenser contains 100 Gem 3 Facet carbon steel single-edge blades. Three facet blades provide an extra-keen edge that enables intricate cutting.