ContactUsLinkComponent
SP Bel-Art Cleanware™ Label Remover, Bel-Art Products, a part of SP
  62866-188
undefined
SP Bel-Art Cleanware™ Label Remover, Bel-Art Products, a part of SP
  62866-188
 :  F17077-0000
  • Description:
    Label-off™ label remover
  • Size:
    59 ml
  • Cat. No.:
    62866-188
  • Supplier no.:
    170770000
Cleanware™ Label-Off™ Label Remover quickly removes labels, including the gummy residue other cleaners leave behind.
  • Quick and easy removal of labels
  • No residue left behind
The kit contains 59 ml (2oz.) of label adhesive solvent in a pump spray with cap, along with 20 lint-free, 10.2 cm (4") square wipes.
 : No air shipment.