Order Entry
Puerto Rico
ContactUsLinkComponent
PIG® Absorb-and-Lock® Acid Absorbent, New Pig
PIG® Absorb-and-Lock® Acid Absorbent, New Pig
Catalog # 89205-864
Supplier:  New Pig
undefined
PIG® Absorb-and-Lock® Acid Absorbent, New Pig
Catalog # 89205-864
Supplier:  New Pig
Supplier Number:  PLP501

Specifications

  • Description:
    Acid Absorbent, Four 4 lb. Containers
  • Cat. No.:
    89205-864
  • Supplier no.:
    PLP501

Specifications

About this item

Permanently solidifies to "lock in" absorbed liquids; won't leak or revert to liquid state when handled or placed under pressure

Forms a solid mass without expanding for easier, less expensive recovery and cleanup; may help you comply with EPA requirements when used to solidify waste prior to shipment or landfilling. High Btu value produces less than 0.1% ash content when burned; ideal for fuels blending and incineration. Shaker bottle makes it easy to apply absorbent directly onto the spill area.

Environmentally friendly product absorbs acids and corrosives including phosphoric, sulfuric, hydrofluoric, hydrochloric and acetic acids as well as hydrazine.