Order Entry
Puerto Rico
ContactUsLinkComponent
PIG® HazMat Chemical Absorbent Dike, New Pig
PIG® HazMat Chemical Absorbent Dike, New Pig
Catalog # 56620-720
Supplier:  New Pig
undefined
PIG® HazMat Chemical Absorbent Dike, New Pig
Catalog # 56620-720
Supplier:  New Pig
Supplier Number:  HA1010

Specifications

  • Capacity:
    Up to 34.1 L (9 gal.) per Sock
  • Description:
    HazMat Chemical Absorbent Dike, 2 Socks
  • Dimensions:
    12.7 cm x 3 m (5" x 10')
  • Cat. No.:
    56620-720
  • Supplier no.:
    HA1010

Specifications

About this item

Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide

Chemical-resistant dike won't degrade or cause a dangerous reaction upon contact with corrosive spills. Excellent for quickly diking and stopping the spread of larger chemical spills; for diking smaller chemical spills, consider our smaller diameter PIG HazMat Socks. Polypropylene skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated. Polypropylene filler is highly absorbent for containing corrosive or reactive spills.