Translucent UN Rated Poly Pail, New Pig
89095-316
: New Pig
- Capacity:18.9 L (5 gal.)
- Dimensions:292×375 mm (11.5×14.75")
- Cat. No.:89095-316
- Supplier no.:DRM282
Translucent walls eliminate guesswork to help prevent overfilling or running out of materials
- Walls eliminate guesswork to help prevent overfilling or running out of materials
- Made from FDA-approved resins for containment of food products
- High-density polyethylene construction is strong and lightweight
- 5-Gallon pail provides efficient storage and transfer of smaller quantities
- Sturdy fold-down handle makes pail easy to carry
- UN1H1/Y1.8/100
Made from FDA Approved resins for containment of food products. High-density polyethylene construction is strong and light-weight. 5-gallon pail provides efficient storage and transfer of smaller quantities. Sturdy fold-down handle makes pail easy to carry. UN1H1/Y1.8/100.