PIG® Traffic Mat® Heavy-Duty Absorbent Rug, New Pig
89205-916
: New Pig
- Description:Heavy weight
- Dimensions:91.4 cmx46 m (36"x150')
- Environmentally Preferable:
- Cat. No.:89205-916
- Supplier no.:MAT162
100% polypropylene material is heat fused and needle punched for incredible durability for walk-on and wheeled traffic
- Absorbs and retains oils, coolants, solvents, and water
- Dark gray color hides grime, so mat stays on the job longer
- Rolls are ideal for walkways and large areas to soak up leaks and overspray
- Cuts easily with scissors or utility knife
- Can be incinerated after use to reduce waste or for fuels blending
- Heavy-duty rug is twice as heavy and absorbent as standard traffic mat rug
Our most durable mat won't rip, shred or fray, even under heavy forklift traffic.