Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant SEMA4A (Ms):Fc (from CHO cells)
Mouse Recombinant SEMA4A (Ms):Fc (from CHO cells)
Catalog # 102511-370
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Mouse Recombinant SEMA4A (Ms):Fc (from CHO cells)
Catalog # 102511-370
Supplier:  Adipogen
Supplier Number:  CHI-MF-111SEMA4-C100
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_038686.3
  • Protein Synonyms:
    Semaphorin-B|Sema B|Sema4a
  • Protein/Peptide Name:
    SEMA4A (Ms):Fc
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102511-370
  • Supplier no.:
    MF111SEMA4C100

Specifications

About this item

Semaphorin 4A (Sema4A, previously semB) is a Class 4 transmembrane Semaphorin with activity in the immune and nervous systems. Sema4A was first described as a molecule that enhances T cell activation and interacts with TIM2 (T cell immunoglobulin and mucin domain 2). Mice with targeted disruption of Sema4A show defects in dendritic cellmediated T cell priming and Th1 responses. Roles for Sema4A have also been identified in the brain, the endothelium and the eye. It mediates hippocampal neuron growth cone collapse in vitro through interaction of the sema domain with plexinB1. Interaction of Sema4A with endothelial cell plexinD1 causes opposition to the angiogenic, proliferative, chemotactic and integrin mediated adhesive actions of VEGF.