Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant SCF (from CHO Cells)
Human Recombinant SCF (from CHO Cells)
Catalog # 102511-364
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Human Recombinant SCF (from CHO Cells)
Catalog # 102511-364
Supplier:  Adipogen
Supplier Number:  CHI-HF-220SCF-C050
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    Shows the biological function of the SCF moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
  • Gene ID:
    NP_003985.2
  • Protein Synonyms:
    c-Kit Ligand|Mast Cell Grow Factor|Stem Cell Factor|MGF|Kit Ligand
  • Protein/Peptide Name:
    SCF
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102511-364
  • Supplier no.:
    HF220SCFC050

Specifications

About this item

Stem cell factor (SCF), also known as cKit ligand (KL), mast cell growth factor (MGF), and steel factor (SLF), is a widely expressed 28-40 kDa type I transmembrane glycoprotein. It promotes the survival, differentiation, and mobilization of multiple cell types including myeloid, erythroid, megakaryocytic, lymphoid, germ cell, and melanocyte progenitors. SCF is a primary growth and activation factor for mast cells and eosinophils. Noncovalent dimers of transmembrane or soluble SCF interact with the receptor tyrosine kinase SCF R/cKit to trigger receptor dimerization and signaling. SCF assists in the recovery of cardiac function following myocardial infarction by increasing the number of cardiomyocytes and vascular channels.