Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant IL21 (from CHO Cells)
Mouse Recombinant IL21 (from CHO Cells)
Catalog # 102513-884
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Mouse Recombinant IL21 (from CHO Cells)
Catalog # 102513-884
Supplier:  Adipogen
Supplier Number:  CHI-MF-12021-C050
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Interleukins
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    Shows the biological function of the IL-21 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
  • Gene ID:
    ABG36530.1
  • Protein Synonyms:
    Interleukin-21
  • Protein/Peptide Name:
    IL21
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102513-884
  • Supplier no.:
    CHIMF12021C050

Specifications

About this item

Interleukin-21 (IL-21) is a key factor in the transition between innate and adaptive immune responses secreted by activated T cells. The IL-21 receptor (IL-21R) is expressed in lymphoid tissue, in particular by NK, B, T and dendritic cells, macrophages and endothelial cells. Recent evidence suggests that IL-21 plays a supportive role in the proliferation of T and B cells and influences the cytolytic activity of natural killer cells. IL-21 has been shown to up-regulate genes associated with innate immunity and to inhibit the differentiation of naïve T helper cells. IL-21 specifically inhibits IFN-gamma production from developing TH1 cells and is preferentially expressed by TH2 cells. Furthermore IL-21 has been identified as a growth and survival factor for human myeloma cells. IL-21/IL-21R interactions have a unique role in sequentially activating both innate and adaptive immune responses against poorly immunogenic tumors, leading to tumor rejection that is perforin dependent but IFN-gamma independent.