About this item
Beauvericin is an antibiotic apoptosis inducer.
- Causes mitochondrial dysfunction
- Cytotoxic
- Anti-cancer compund
Antibiotic. Apoptosis inducer. Acyl-CoA:cholesterol acyltransferase inhibitor. Anticancer compound. Antihaptotactic and antimetastatic. Antiangiogenic compound. Antibacterial, antiprotozal, antiviral and antifungal compound. Shows ionophoric properties. Cytotoxic. Genotoxic. Potently interacts with ABCB1 and ABCG2 transport functions.
White to off-white solid.
Stable for at least two years after receipt when stored at –20 °C. Store solutions at –20 °C in the dark.
Soluble in ethanol, methanol or DMSO.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:26048-05-5
- Storage Conditions:Freezer
- Protein/Peptide Name:Beauvericin
- Purity:97% (HPLC assay method)
- Molecular Weight:784.0 g/mol
- Molecular Formula:C₄₅H₅₇N₃O₉