Order Entry
Puerto Rico
ContactUsLinkComponent
Phorbol 12-myristate 13-acetate ≥98% (by HPLC)
Irritant
Phorbol 12-myristate 13-acetate ≥98% (by HPLC)
Supplier:  Adipogen
Certificates
Irritant
Phorbol 12-myristate 13-acetate ≥98% (by HPLC)
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Source/Host:
    Semisynthetic.
  • Purity:
    98% (HPLC)
  • Appearance/Color:
    White to off-white solid.
  • Solubility:
    Soluble in DMSO, methanol, 100% ethanol, acetone, ether or dimethylformamide; almost insoluble in aqueous buffers.

Specification Test Results

Frequently Bought Together