Human Recombinant BCMA (from HEK293 cells)
: Adipogen
BCMA is a receptor for BAFF and APRIL. It promotes B cell survival and plays a role in the regulation of humoral immunity. Its downstream signaling is dependent on NF-kappaB and JNK.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.01EU/µg
- Biological Activity:Blocks the binding of BAFF and APRIL to their receptors BCMA and TACI, inhibiting BAFF- and APRIL-mediated B cell activation.
- Gene ID:Q02223
- Protein Synonyms:B Cell Maturation Protein|CD269|TNFRSF17
- Protein/Peptide Name:BCMA
- Purity:95%
- Molecular Weight:~40kDa (SDS-PAGE)
- Sequence:19756
- Endotoxin Level:Low
- Concentration:1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping Temperature:-20 °C, Blue Ice