About this item
FGF-21 (Fibroblast growth factor 21) stimulates glucose uptake in differentiated adipocytes via the induction of glucose transporter SLC2A1/GLUT1 expression (but not SLC2A4/GLUT4 expression). Activity requires the presence of betaKlotho.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:<5EU/mg
- Gene ID:Q9JJN1
- Protein Synonyms:Fibroblast Growth Factor 21
- Protein/Peptide Name:FGF21
- Purity:90%
- Molecular Weight:~50kDa (SDS-PAGE)
- Sequence:1-210
- Endotoxin Level:Low
- Formulation:Liquid. 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice